SY3H3D6790D4 Synthonix Corporation

(S)-Benzyl 6-(hydroxymethyl)-5,6-dihydropyridine-1(2H)-carboxylate

  • MDL number MFCD19703949
  • Cas Number : 1808097-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57744.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57744.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart