SY3H3D6790D7 Synthonix Corporation

2-(tert-Butoxycarbonyl)isoindoline-1-carboxylic acid

  • MDL number MFCD01321007
  • Cas Number : 221352-46-1
  • Molecular Weight : 263.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57753.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57753.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart