SY3H3D6790E6 Synthonix Corporation

(R)-1-Benzyl 2-methyl piperazine-1,2-dicarboxylate

  • MDL number MFCD04115343
  • Cas Number : 1217598-28-1
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57776.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57776.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart