SY3H3D6790F4 Synthonix Corporation

3-tert-Butoxycarbonylamino-3-(3-chlorophenyl)propionic acid

  • MDL number MFCD02090710
  • Cas Number : 284493-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57799.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57799.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart