SY3H3D6790FB Synthonix Corporation

Benzyl tert-butyl malonate

  • MDL number MFCD01075175
  • Cas Number : 72594-86-6
  • Molecular Weight : 250.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzyl 3-tert-butyl malonate
Inchi Code 1S/C14H18O4/c1-14(2,3)18-13(16)9-12(15)17-10-11-7-5-4-6-8-11/h4-8H,9-10H2,1-3H3
Inchi Key XKXXXODAXXAFNP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Light yellow to yellow liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/57817.html

Safety Information

signalword Warning
pcodes P280,P305,P351,P338
MSDS https://synthonix.com/57817.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart