SY3H3D679117 Synthonix Corporation

Boc-(2R,3S)-3-Phenylisoserine

  • Cas Number : 145514-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57866.html

Safety Information

MSDS https://synthonix.com/57866.html
Availability details for
Quantity
  • Availability
  • Add to Cart