SY3H3D679118 Synthonix Corporation

trans-2-(4-Bromophenyl)-5-propyltetrahydro-2H-pyran

  • MDL number MFCD30536236
  • Cas Number : 700863-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57876.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57876.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart