SY3H3D67911F Synthonix Corporation

(S)-Benzyl (1-amino-4-methyl-1-oxopentan-2-yl)carbamate

  • MDL number MFCD00080252
  • Cas Number : 4801-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57889.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57889.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart