SY3H3D6799E8 Synthonix Corporation

O-(Carboxymethyl)hydroxylamine hemihydrochloride

  • MDL number MFCD00012955
  • Cas Number : 2921-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40302.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40302.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart