SY3H3D6799E9 Synthonix Corporation

2-(Chloromethyl)piperidine hydrochloride

  • MDL number MFCD02667639
  • Cas Number : 2554-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41543.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41543.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart