SY3H3D6799EC Synthonix Corporation

2-tert-Butyl 4-ethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate

  • MDL number MFCD02179394
  • Cas Number : 86770-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57933.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57933.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart