SY3H3D6799FC Synthonix Corporation

(2R,6S)-rel-tert-Butyl 2,6-diethyl-4-oxopiperidine-1-carboxylate

  • Cas Number : 1003843-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57987.html

Safety Information

MSDS https://synthonix.com/57987.html
Availability details for
Quantity
  • Availability
  • Add to Cart