SY3H3D679A0E Synthonix Corporation

2-Bromoanthracene-9,10-dione

  • MDL number MFCD00474541
  • Cas Number : 572-83-8
  • Molecular Weight : 287.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Pale yellow - Greyish reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58024.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58024.html
pictograms GHS07
hstatements H302, H315, H319
Availability details for
Quantity
  • Availability
  • Add to Cart