SY3H3D679A18 Synthonix Corporation

(6-Bromoimidazo[1,2-a]pyridin-3-yl)(phenyl)methanone

  • Cas Number : 1284293-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58050.html

Safety Information

MSDS https://synthonix.com/58050.html
Availability details for
Quantity
  • Availability
  • Add to Cart