SY3H3D679A1A Synthonix Corporation

Benzo[h]quinoline-2-carbaldehyde

  • Cas Number : 904886-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58055.html

Safety Information

MSDS https://synthonix.com/58055.html
Availability details for
Quantity
  • Availability
  • Add to Cart