SY3H3D679A2B Synthonix Corporation

2-Bromo-4,6-diphenyl-1,3,5-triazine

  • Cas Number : 80984-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58113.html

Safety Information

MSDS https://synthonix.com/58113.html
Availability details for
Quantity
  • Availability
  • Add to Cart