SY3H3D679A2E Synthonix Corporation

9-(Bromomethyl)anthracene

  • Cas Number : 2417-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58128.html

Safety Information

MSDS https://synthonix.com/58128.html
Availability details for
Quantity
  • Availability
  • Add to Cart