SY3H3D679A33 Synthonix Corporation

1-Benzyl-1H-indazole-3-carboxylic acid

  • Cas Number : 41354-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58150.html

Safety Information

MSDS https://synthonix.com/58150.html
Availability details for
Quantity
  • Availability
  • Add to Cart