SY3H3D679A3C Synthonix Corporation

4-Bromo-9,9-dimethyl-9H-fluorene

  • MDL number MFCD23099415
  • Cas Number : 942615-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Pale-yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58171.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58171.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart