SY3H3D679A3D Synthonix Corporation

Benzyl 2,4-dichloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate

  • MDL number MFCD19707615
  • Cas Number : 1370411-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58178.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58178.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart