SY3H3D679A41 Synthonix Corporation

2-Benzamido-2-phenylacetic acid

  • MDL number MFCD00666894
  • Cas Number : 74536-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58197.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58197.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart