SY3H3D679A4D Synthonix Corporation

N-Benzyl-4-bromo-3-methylbenzamide

  • MDL number MFCD09972121
  • Cas Number : 1020252-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58223.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58223.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart