SY3H3D679A60 Synthonix Corporation

1-Benzyl 3-methyl 4-oxopiperidine-1,3-dicarboxylate

  • Cas Number : 159299-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58304.html

Safety Information

MSDS https://synthonix.com/58304.html
Availability details for
Quantity
  • Availability
  • Add to Cart