SY3H3D679A63 Synthonix Corporation

tert-Butyl (2-methylquinolin-3-yl)carbamate

  • MDL number MFCD19689605
  • Cas Number : 1824282-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58311.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58311.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart