SY3H3D679A67 Synthonix Corporation

tert-Butyl 2-(6-bromo-2-oxo-3,4-dihydroquinolin-1(2H)-yl)acetate

  • MDL number MFCD26392965
  • Cas Number : 1224927-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58322.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58322.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart