SY3H3D679A70 Synthonix Corporation

(3S,4R)-1-tert-Butyl-4-(2,4-difluorophenyl)pyrrolidine-3-carboxylic acid

  • Cas Number : 455957-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58344.html

Safety Information

MSDS https://synthonix.com/58344.html
Availability details for
Quantity
  • Availability
  • Add to Cart