SY3H3D679AD9 Synthonix Corporation

(R)-3-((tert-Butoxycarbonyl)amino)-4-(4-iodophenyl)butanoic acid

  • MDL number MFCD01860963
  • Cas Number : 269396-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58380.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58380.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart