SY3H3D679ADA Synthonix Corporation

1-Bromo-4-(trans-4-propylcyclohexyl)benzene

  • MDL number MFCD00577367
  • Cas Number : 86579-53-5
  • Molecular Weight : 281.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-4-(4-propylcyclohexyl)benzene
Inchi Code 1S/C15H21Br/c1-2-3-12-4-6-13(7-5-12)14-8-10-15(16)11-9-14/h8-13H,2-7H2,1H3/t12-,13-
Inchi Key GMZABADCQVWQPS-JOCQHMNTSA-N

Description

Purity 96%
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/58386.html

Safety Information

signalword Warning
pcodes P280,P305,P351,P338
MSDS https://synthonix.com/58386.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart