SY3H3D679ADC Synthonix Corporation

tert-Butyl 2-(4-chlorophenyl)piperazine-1-carboxylate

  • MDL number MFCD08275679
  • Cas Number : 769944-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid or Semi-solid or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58390.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58390.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart