SY3H3D679ADF Synthonix Corporation

tert-Butyl 3-benzylazetidine-1-carboxylate

  • MDL number MFCD30723490
  • Cas Number : 1714979-63-1
  • Molecular Weight : 247.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58396.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58396.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart