SY3H3D679AE1 Synthonix Corporation

tert-Butyl ((R)-1-((S)-oxiran-2-yl)-2-phenylethyl)carbamate

  • MDL number MFCD08061626
  • Cas Number : 156474-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58400.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58400.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart