SY3H3D679AE4 Synthonix Corporation

Boc-D-beta-Homophenylalanine

  • MDL number MFCD01076270
  • Cas Number : 101555-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form powder Colour: White - off-white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58403.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58403.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart