SY3H3D679AF3 Synthonix Corporation

tert-Butyl 4-(6-bromopyridin-2-yl)-1,4-diazepane-1-carboxylate

  • Cas Number : 1152093-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58435.html

Safety Information

MSDS https://synthonix.com/58435.html
Availability details for
Quantity
  • Availability
  • Add to Cart