SY3H3D679B01 Synthonix Corporation

(S)-1-((S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carbonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00057246
  • Cas Number : 15401-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58475.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58475.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart