SY3H3D679B0C Synthonix Corporation

(S)-tert-Butyl 3-((tert-butoxycarbonyl)amino)piperidine-1-carboxylate

  • MDL number MFCD09701971
  • Cas Number : 1217710-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58504.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58504.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart