SY3H3D679B15 Synthonix Corporation

tert-Butyl 3-aminopyrrolidine-1-carboxylate 2-hydroxypropane-1,2,3-tricarboxylate

  • MDL number MFCD16294163
  • Cas Number : 1310278-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58534.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58534.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart