SY3H3D679B19 Synthonix Corporation

tert-Butyl 2-(3-(aminomethyl)phenyl)acetate oxalate

  • Cas Number : 932740-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58547.html

Safety Information

MSDS https://synthonix.com/58547.html
Availability details for
Quantity
  • Availability
  • Add to Cart