SY3H3D679B1C Synthonix Corporation

2-Benzoylbenzofuran-5-carbaldehyde

  • Cas Number : 120973-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58554.html

Safety Information

MSDS https://synthonix.com/58554.html
Availability details for
Quantity
  • Availability
  • Add to Cart