SY3H3D679B1D Synthonix Corporation

[1,1'-Biphenyl]-3,3',4,4'-tetracarboxylic acid

  • Cas Number : 22803-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58556.html

Safety Information

MSDS https://synthonix.com/58556.html
Availability details for
Quantity
  • Availability
  • Add to Cart