SY3H3D679B21 Synthonix Corporation

2-(4-Bromophenyl)-5-phenylthiophene

  • Cas Number : 118621-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58560.html

Safety Information

MSDS https://synthonix.com/58560.html
Availability details for
Quantity
  • Availability
  • Add to Cart