SY3H3D679B24 Synthonix Corporation

5-Bromo-2,3-diphenylpyrazine

  • MDL number MFCD00234886
  • Cas Number : 243472-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58563.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58563.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart