SY3H3D679B25 Synthonix Corporation

4-Bromo-2,6-diphenylpyrimidine

  • MDL number MFCD00234885
  • Cas Number : 40734-24-5
  • Molecular Weight : 311.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58564.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58564.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart