SY3H3D679B26 Synthonix Corporation

3-Bromo-5-(2-pyridyl)-1-phenyl-1,2-dihydropyridin-2-one

  • MDL number MFCD21607392
  • Cas Number : 381248-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Off-white to Yellow to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58571.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58571.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart