SY3H3D679B2A Synthonix Corporation

7-(Benzyloxy)-2,4-dichloro-6-methoxyquinazoline

  • Cas Number : 60771-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58586.html

Safety Information

MSDS https://synthonix.com/58586.html
Availability details for
Quantity
  • Availability
  • Add to Cart