SY3H3D679B31 Synthonix Corporation

5-Benzyloxyindole-3-carboxaldehyde

  • MDL number MFCD00014562
  • Cas Number : 6953-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form powder Colour: white - yellow or light grey
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58608.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58608.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart