SY3H3D679B37 Synthonix Corporation

1-(3-(benzyloxy)-2-fluoro-6-nitrophenyl)propan-2-one

  • MDL number MFCD09838836
  • Cas Number : 288385-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58633.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58633.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart