SY3H3D679B39 Synthonix Corporation

(E)-3-(4-(Benzyloxy)phenyl)acrylic acid

  • MDL number MFCD00021778
  • Cas Number : 6272-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58642.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58642.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart