SY3H3D679B43 Synthonix Corporation

4-(Benzyloxy)-3,5-dimethoxybenzaldehyde

  • Cas Number : 6527-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58685.html

Safety Information

MSDS https://synthonix.com/58685.html
Availability details for
Quantity
  • Availability
  • Add to Cart