SY3H3D679B45 Synthonix Corporation

4-Butyl-4'-iodobiphenyl

  • MDL number MFCD16619281
  • Cas Number : 199982-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58690.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58690.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart