SY3H3D679B4C Synthonix Corporation

2-(Benzyl(methyl)amino)-1-phenylethanol

  • MDL number MFCD01652783
  • Cas Number : 29194-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58723.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58723.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart